Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRIP3
Entrez Gene ID
401262
Gene Name
cysteine rich protein 3
Gene Aliases
TLP, TLP-A, bA480N24.2
Chromosome Location
Chr.6: 43305473 - 43310705 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005249103.3 XP_005249160.1 1 - 1 1745 100
XM_011514609.2 XP_011512911.1 1 - 1 1745 100
XM_017010856.1 XP_016866345.1 1 - 1 1745 100
XM_017010857.1 XP_016866346.1 1 - 1 1745 100


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX952027.1 - 1 291 Chr. 6 - 43308689 - 43309251 [-] on Build GRCh38 100
GenBankmRNA AK124657.1 - 2 1148 Chr. 6 - 43307043 - 43310883 [-] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream solute carrier family 22 member 7 SLC22A7 NLT, OAT2 -2.4kb Anti-sense Chr. - 43295694 - 43305538 on Build GRCh38
Downstream zinc finger protein 318 ZNF318 HRIHFB2436, TZF, ZFP318 17.6kb Sense Chr. - 43327489 - 43369676 on Build GRCh38

View Full Product Details