Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TXNRD1
Entrez Gene ID
7296
Gene Name
thioredoxin reductase 1
Gene Aliases
GRIM-12, TR, TR1, TRXR1, TXNR
Chromosome Location
Chr.12: 104215779 - 104350307 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY423378.1 - 5 - 5 445 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream nuclear transcription factor Y subunit beta NFYB CBF-A, CBF-B, HAP3, NF-YB -146.0kb Anti-sense Chr. - 104117080 - 104138262 on Build GRCh38
Downstream EP300 interacting inhibitor of differentiation 3 EID3 NS4EB, NSE4B, NSMCE4B 19.3kb Sense Chr. - 104303732 - 104305205 on Build GRCh38

View Full Product Details