Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ANK3
- Entrez Gene ID
-
288
- Gene Name
- ankyrin 3, node of Ranvier (ankyrin G)
- Gene Aliases
-
ANKYRIN-G, MRT37
- Chromosome Location
-
Chr.10: 60026298 - 60733526 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK172736.1 |
- |
3 - 3 |
644 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 1553 |
LINC01553 |
C10orf40 |
-546.6kb
|
Sense |
Chr. - 59958217 - 59960913 on Build GRCh38 |
Downstream |
cyclin dependent kinase 1 |
CDK1 |
CDC2, CDC28A, P34CDC2 |
269.7kb
|
Anti-sense |
Chr. - 60778331 - 60794852 on Build GRCh38 |
View Full Product Details