The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001720.3 | NP_001711.2 | 7 - 7 | 1548 | 72 |
XM_011542024.2 | XP_011540326.2 | 8 - 8 | 1341 | 72 | |
GenBank mRNA | AY303955.1 | - | 7 - 7 | 1548 | 72 |
BC023526.2 | - | 7 - 7 | 1147 | 72 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181809.3 | NP_861525.2 | 7 - 7 | 1518 | 72 |
GenBank mRNA | AY303954.1 | - | 7 - 7 | 1518 | 72 |
BC131617.1 | - | 7 - 7 | 834 | 72 |