Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TYRO3
Entrez Gene ID
7301
Gene Name
TYRO3 protein tyrosine kinase
Gene Aliases
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif
Chromosome Location
Chr.15: 41559022 - 41579338 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006293.3 NP_006284.2 5 - 6 906 67
XM_017022542.1 XP_016878031.1 5 - 6 871 67
XM_017022543.1 XP_016878032.1 5 - 6 707 67
GenBank mRNA AB209431.1 - 5 - 6 935 67
BC034960.1 - 5 - 6 688 67
BC049368.1 - 5 - 6 680 67
BC051756.1 - 5 - 6 685 67
D17517.1 - 5 - 6 894 67
D50479.1 - 5 - 6 666 67
U02566.1 - 2 - 3 117 67
U05682.1 - 5 - 6 676 67

View Full Product Details