The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000191.2 | NP_000182.2 | 8 - 9 | 921 | 88 |
NM_001166059.1 | NP_001159531.1 | 6 - 7 | 708 | 88 | |
GenBank mRNA | AK122801.1 | - | 9 - 10 | 1474 | 88 |
AK300733.1 | - | 6 - 7 | 677 | 88 | |
AK313869.1 | - | 8 - 9 | 890 | 88 | |
BC010570.1 | - | 8 - 9 | 887 | 88 | |
BT009792.1 | - | 8 - 9 | 877 | 88 | |
CR456884.1 | - | 8 - 9 | 877 | 88 | |
L07033.1 | - | 8 - 9 | 891 | 88 |