The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173588.3 | NP_775859.3 | 17 - 18 | 2895 | 79 |
XM_011520030.2 | XP_011518332.1 | 15 - 16 | 5799 | 79 | |
XM_011520031.2 | XP_011518333.1 | 14 - 15 | 5496 | 79 | |
XM_017017607.1 | XP_016873096.1 | 16 - 17 | 5690 | 79 | |
XM_017017608.1 | XP_016873097.1 | 16 - 17 | 3713 | 79 | |
GenBank mRNA | AK095113.1 | - | 16 - 17 | 2592 | 79 |
BC115396.1 | - | 17 - 18 | 2846 | 79 | |
BC115397.1 | - | 16 - 17 | 3045 | 79 |