Product Details

Assay Reference Genome
Location

Chr.19:46752356 on build GRCh38
Cytoband
19q13.32
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
79147
Gene Symbol
FKRP
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
Location
Chr.19:46746015-46758575 on build GRCh38
Assay Gene Location
Within Intron 4
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
FKRP NM_001039885.2 NP_001034974.1
NM_024301.4 NP_077277.1
XM_005259247.2 XP_005259304.1
XM_005259248.2 XP_005259305.1
XM_005259249.4 XP_005259306.1
XM_011527306.1 XP_011525608.1
XM_011527307.1 XP_011525609.1
XM_017027297.1 XP_016882786.1
AJ314847.1 CAC85633.1
AK022638.1 BAB14146.1
AK095497.1
AK291282.1
BC002612.2 AAH02612.1
BM979509.1
DA123579.1
DB090073.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833850 Chr19:46703285 - 46867053 on Build GRCh38 Loss PRKD2 MIR320E ARHGAP35 SNAR-E FKRP AP2S1 STRN4 LOC107987269 SLC1A5

View Full Product Details