Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBTFL1
Entrez Gene ID
642623
Gene Name
upstream binding transcription factor, RNA polymerase I-like 1
Gene Aliases
C11orf27, HMGPI
Chromosome Location
Chr.11: 90085950 - 90087131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143975.1 NP_001137447.1 1 - 1 237 92

View Full Product Details