Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDFIC
Entrez Gene ID
29969
Gene Name
MyoD family inhibitor domain containing
Gene Aliases
HIC
Chromosome Location
Chr.7: 114922154 - 115019916 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166345.1 NP_001159817.1 1 - 2 480 113
NM_001166346.1 NP_001159818.3 1 - 2 480 113
NM_199072.4 NP_951038.1 1 - 2 480 113
GenBank mRNA AF054589.1 - 1 - 2 480 113

View Full Product Details