The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009998.3 | NP_001009998.1 | 14 - 15 | 1239 | 55 |
NM_032627.4 | NP_116016.1 | 15 - 16 | 1305 | 55 | |
XM_005259790.3 | XP_005259847.1 | 15 - 16 | 1346 | 55 | |
XM_006722665.3 | XP_006722728.1 | 15 - 16 | 1352 | 55 | |
XM_006722666.2 | XP_006722729.1 | 15 - 16 | 1310 | 55 | |
XM_006722668.2 | XP_006722731.1 | 14 - 15 | 1244 | 55 | |
XM_017026431.1 | XP_016881920.1 | 15 - 16 | 1364 | 55 | |
XM_017026432.1 | XP_016881921.1 | 15 - 16 | 1902 | 55 | |
XM_017026433.1 | XP_016881922.1 | 15 - 16 | 1310 | 55 | |
XM_017026434.1 | XP_016881923.1 | 14 - 15 | 1293 | 55 | |
XM_017026435.1 | XP_016881924.1 | 14 - 15 | 1835 | 55 | |
XM_017026436.1 | XP_016881925.1 | 14 - 15 | 1244 | 55 | |
XM_017026437.1 | XP_016881926.1 | 14 - 15 | 1275 | 55 | |
GenBank mRNA | AK098429.1 | - | 9 - 10 | 820 | 55 |
BC000274.1 | - | 15 - 16 | 1212 | 55 |