The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032387.1 | NP_001027559.1 | 1 - 2 | 82 | 77 |
GenBank mRNA | AK312763.1 | - | 1 - 2 | 56 | 77 |
AU126450.1 | - | 1 - 2 | 82 | 77 |