The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114635.1 | NP_001108107.1 | 2 - 3 | 280 | 85 |
XM_017013577.1 | XP_016869066.1 | 3 - 4 | 412 | 85 | |
GenBank mRNA | AK296933.1 | - | 2 - 3 | 280 | 85 |
DC347559.1 | - | 2 - 3 | 280 | 85 |