Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BRD7
Entrez Gene ID
29117
Gene Name
bromodomain containing 7
Gene Aliases
BP75, CELTIX1, NAG4
Chromosome Location
Chr.16: 50317563 - 50368969 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173984.2 NP_001167455.1 5 - 6 742 106
NM_013263.4 NP_037395.2 5 - 6 742 106
XM_011523046.2 XP_011521348.1 5 - 6 828 106
XM_011523047.2 XP_011521349.1 5 - 6 828 106
XM_011523048.2 XP_011521350.1 5 - 6 828 106
XM_011523049.2 XP_011521351.1 5 - 6 828 106
XM_017023179.1 XP_016878668.1 5 - 6 642 106
XM_017023180.1 XP_016878669.1 5 - 6 642 106
GenBank mRNA AF152604.1 - 5 - 6 635 106
AF213969.1 - 5 - 6 587 106
AJ271881.1 - 5 - 6 700 106
BC050728.1 - 6 - 7 649 106
BC094706.1 - 6 - 7 619 106

View Full Product Details