The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003708.4 | NP_003699.3 | 2 - 3 | 2485 | 91 |
GenBank mRNA | AF057034.1 | - | 2 - 3 | 709 | 91 |
AF086735.1 | - | 2 - 3 | 1425 | 91 |