The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161416.1 | NP_001154888.1 | 2 - 3 | 612 | 78 |
GenBank mRNA | DA366480.1 | - | 2 - 3 | 484 | 78 |