Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GPR17
Entrez Gene ID
2840
Gene Name
G protein-coupled receptor 17
Gene Aliases
-
Chromosome Location
Chr.2: 127645864 - 127652639 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161417.1 NP_001154889.1 1 - 2 378 74
GenBank mRNA AK292725.1 - 1 - 2 248 74
BC031653.1 - 1 - 2 57 74
BX538082.1 - 1 - 2 192 74
DA154324.1 - 1 - 2 378 74
DA390892.1 - 1 - 2 357 74

View Full Product Details