The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019054.2 | NP_061927.2 | 2 - 3 | 111 | 97 |
XM_005269920.3 | XP_005269977.1 | 2 - 3 | 101 | 97 | |
XM_006717903.3 | XP_006717966.1 | 2 - 3 | 101 | 97 | |
XM_017016350.1 | XP_016871839.1 | 2 - 3 | 369 | 97 | |
XM_017016351.1 | XP_016871840.1 | 2 - 3 | 101 | 97 | |
XM_017016352.1 | XP_016871841.1 | 2 - 3 | 101 | 97 | |
XM_017016353.1 | XP_016871842.1 | 2 - 3 | 101 | 97 | |
GenBank mRNA | AK022978.1 | - | 2 - 3 | 111 | 97 |
BC051863.2 | - | 1 - 2 | 57 | 97 |