The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024070.3 | NP_076975.2 | 1 - 2 | 155 | 84 |
XM_011516575.2 | XP_011514877.1 | 2 - 3 | 171 | 84 | |
GenBank mRNA | BC107873.1 | - | 2 - 3 | 180 | 84 |