The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145212.3 | NP_660213.1 | 5 - 6 | 556 | 104 |
NR_028356.1 | 5 - 6 | 556 | 104 | ||
GenBank mRNA | AK025547.1 | - | 5 - 6 | 484 | 104 |
AK225894.1 | - | 5 - 6 | 452 | 104 | |
BC000217.1 | - | 5 - 6 | 446 | 104 | |
BC022391.1 | - | 5 - 6 | 520 | 104 | |
BG532984.1 | - | 5 - 6 | 438 | 104 | |
BU154006.1 | - | - | 429 | 104 | |
BU533068.1 | - | 5 - 6 | 486 | 104 |