The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016334.3 | NP_057418.1 | 7 - 8 | 762 | 102 |
XM_005277401.3 | XP_005277458.1 | 7 - 8 | 735 | 102 | |
XM_005277402.4 | XP_005277459.1 | 7 - 8 | 899 | 102 | |
XM_005277404.3 | XP_005277461.1 | 7 - 8 | 755 | 102 | |
XM_006711377.3 | XP_006711440.1 | 8 - 9 | 798 | 102 | |
XM_006711378.2 | XP_006711441.1 | 8 - 9 | 801 | 102 | |
XM_011509610.2 | XP_011507912.1 | 8 - 9 | 795 | 102 | |
XM_011509612.2 | XP_011507914.1 | 8 - 9 | 756 | 102 | |
XM_011509613.2 | XP_011507915.1 | 7 - 8 | 735 | 102 | |
XM_017001448.1 | XP_016856937.1 | 8 - 9 | 1298 | 102 | |
XM_017001449.1 | XP_016856938.1 | 7 - 8 | 735 | 102 | |
GenBank mRNA | AK299995.1 | - | 7 - 8 | 733 | 102 |
AK302266.1 | - | 7 - 8 | 754 | 102 | |
AL133020.1 | - | 7 - 8 | 746 | 102 | |
CD102528.1 | - | 2 - 3 | 127 | 102 | |
KJ534849.1 | - | 7 - 8 | 618 | 102 | |
KJ534850.1 | - | 7 - 8 | 618 | 102 | |
KJ534851.1 | - | 7 - 8 | 549 | 102 | |
KJ535070.1 | - | 8 - 9 | 946 | 102 | |
U78723.1 | - | 7 - 8 | 721 | 102 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001097612.1 | NP_001091081.1 | 7 - 8 | 792 | 102 |
NM_001097613.2 | NP_001091082.2 | 8 - 9 | 838 | 102 | |
NR_036541.1 | 7 - 8 | 812 | 102 | ||
XM_006711492.3 | XP_006711555.1 | 7 - 8 | 762 | 102 | |
XM_006711493.3 | XP_006711556.1 | 8 - 9 | 770 | 102 | |
XM_011509908.2 | XP_011508210.1 | 8 - 9 | 808 | 102 | |
XM_011509909.2 | XP_011508211.1 | 7 - 8 | 1020 | 102 | |
XM_011509910.2 | XP_011508212.1 | 7 - 8 | 762 | 102 | |
XM_017002150.1 | XP_016857639.1 | 7 - 8 | 762 | 102 | |
GenBank mRNA | AB097024.1 | - | 7 - 8 | 733 | 102 |
AB097025.1 | - | 7 - 8 | 733 | 102 | |
AF132947.1 | - | 7 - 8 | 717 | 102 | |
AK021758.1 | - | 7 - 8 | 782 | 102 | |
AK223223.1 | - | 7 - 8 | 733 | 102 | |
AK301939.1 | - | 7 - 8 | 639 | 102 | |
AK316371.1 | - | 8 - 9 | 750 | 102 | |
AY358966.1 | - | 7 - 8 | 767 | 102 | |
BC003187.1 | - | 7 - 8 | 695 | 102 | |
BC067816.1 | - | 7 - 8 | 751 | 102 | |
BC146880.1 | - | 7 - 8 | 713 | 102 | |
BT006679.1 | - | 7 - 8 | 618 | 102 | |
KJ534847.1 | - | 7 - 8 | 618 | 102 | |
KJ534848.1 | - | 7 - 8 | 618 | 102 | |
KJ535029.1 | - | 7 - 8 | 618 | 102 | |
KJ535049.1 | - | 7 - 8 | 638 | 102 |