Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TBC1D3B
- Entrez Gene ID
-
414059
- Gene Name
- TBC1 domain family member 3B
- Gene Aliases
-
PRC17, TBC1D3I, TBC1D3L
- Chromosome Location
-
Chr.17: 36165681 - 36176624 on Build GRCh38
- Gene Symbol
- TBC1D3C
- Entrez Gene ID
-
414060
- Gene Name
- TBC1 domain family member 3C
- Gene Aliases
-
TBC1D3D
- Chromosome Location
-
Chr.17: 38057693 - 38068634 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK313329.1 |
- |
6 - 7 |
523 |
92 |
- Gene Symbol
- TBC1D3E
- Entrez Gene ID
-
102723859
- Gene Name
- TBC1 domain family member 3E
- Gene Aliases
-
TBC1DE3
- Chromosome Location
-
Chr.17: 38119293 - 38138868 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC047739.1 |
- |
7 - 8 |
694 |
92 |
- Gene Symbol
- TBC1D3F
- Entrez Gene ID
-
84218
- Gene Name
- TBC1 domain family member 3F
- Gene Aliases
-
TBC1D3
- Chromosome Location
-
Chr.17: 36428612 - 36439523 on Build GRCh38
- Gene Symbol
- TBC1D3G
- Entrez Gene ID
-
101060321
- Gene Name
- TBC1 domain family member 3G
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 36322242 - 36334759 on Build GRCh38
- Gene Symbol
- TBC1D3K
- Entrez Gene ID
-
101060351
- Gene Name
- TBC1 domain family member 3K
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 37923805 - 37935371 on Build GRCh38
View Full Product Details