Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SFTPA1
Entrez Gene ID
653509
Gene Name
surfactant protein A1
Gene Aliases
COLEC4, PSAP, PSP-A, PSPA, SFTP1, SFTPA1B, SP-A, SP-A1, SPA, SPA1
Chromosome Location
Chr.10: 79610939 - 79615443 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001093770.2 NP_001087239.2 6 - 6 1777 102
NM_001164644.1 NP_001158116.1 6 - 6 1747 102
NM_001164645.1 NP_001158117.1 6 - 6 1633 102
NM_001164646.1 NP_001158118.1 6 - 6 1641 102
NM_001164647.1 NP_001158119.1 5 - 5 1717 102
NM_005411.4 NP_005402.3 6 - 6 1788 102
XM_005270062.4 XP_005270119.1 5 - 5 1777 102
XM_006717953.2 XP_006718016.1 6 - 6 1770 102
GenBank mRNA BC026229.1 - 2 - 2 1260 102
HQ021433.2 - 5 - 5 1717 102
HQ021434.1 - 5 - 5 1728 102
HQ021435.1 - 5 - 5 1717 102
HQ021436.1 - 5 - 5 1716 102
HQ021437.1 - 6 - 6 1799 102
HQ021438.1 - 6 - 6 1788 102
HQ021439.1 - 6 - 6 1787 102
HQ021440.1 - 6 - 6 1788 102
HQ021441.1 - 6 - 6 1777 102
HQ021442.1 - 6 - 6 1776 102
JX502764.1 - 6 - 6 1788 102
JX502765.1 - 6 - 6 1777 102

View Full Product Details