The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032834.3 | NP_116223.3 | 1 - 1 | 153 | 89 |
GenBank mRNA | AJ312278.1 | - | 1 - 1 | 153 | 89 |