Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM71E1
Entrez Gene ID
112703
Gene Name
family with sequence similarity 71 member E1
Gene Aliases
-
Chromosome Location
Chr.19: 50466643 - 50476746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308429.1 NP_001295358.1 1 - 1 49 82
NM_138411.2 NP_612420.1 1 - 1 49 82
GenBank mRNA BC071737.1 - 1 - 1 49 82

View Full Product Details