Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP4A11
Entrez Gene ID
1579
Gene Name
cytochrome P450 family 4 subfamily A member 11
Gene Aliases
CP4Y, CYP4A2, CYP4AII
Chromosome Location
Chr.1: 46929174 - 46941484 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000778.3 NP_000769.2 12 - 12 2352 114
NM_001319155.1 NP_001306084.1 12 - 12 2256 114
NR_134988.1 10 - 10 2006 114
NR_134989.1 11 - 11 2197 114
NR_134990.1 11 - 11 2191 114
NR_134991.1 11 - 11 2178 114
NR_134992.1 8 - 8 1807 114
NR_134993.1 9 - 9 1941 114
NR_134994.1 11 - 11 2213 114
XM_017000464.1 XP_016855953.1 10 - 10 2050 114
XM_017000465.1 XP_016855954.1 12 - 12 2271 114
GenBank mRNA D26481.1 - 12 - 12 2315 114
L04751.1 - 12 - 12 2336 114
S67580.1 - 12 - 12 2343 114
S67581.1 - 12 - 12 2342 114

Gene Symbol
CYP4A22
Entrez Gene ID
284541
Gene Name
cytochrome P450 family 4 subfamily A member 22
Gene Aliases
-
Chromosome Location
Chr.1: 47137425 - 47149738 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270767.3 XP_005270824.1 10 - 10 2053 114

View Full Product Details