The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242362.1 | NP_001229291.1 | 1 - 1 | 49 | 90 |
NM_030801.3 | NP_110428.2 | 1 - 1 | 49 | 90 | |
NM_177535.2 | NP_803879.1 | 1 - 1 | 49 | 90 | |
NM_177537.2 | NP_803881.1 | 1 - 1 | 49 | 90 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098800.2 | NP_001092270.1 | 1 - 1 | 49 | 90 |
NM_001272061.1 | NP_001258990.1 | 1 - 1 | 49 | 90 | |
NM_001272062.1 | NP_001258991.1 | 1 - 1 | 49 | 90 | |
NM_001272063.1 | NP_001258992.1 | 1 - 1 | 49 | 90 |