The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105549.1 | NP_001099019.1 | 6 - 6 | 735 | 88 |
NM_001105550.1 | NP_001099020.1 | 5 - 5 | 619 | 88 | |
NM_001105551.1 | NP_001099021.1 | 5 - 5 | 645 | 88 | |
NM_001105552.1 | NP_001099022.1 | 4 - 4 | 521 | 88 | |
NM_001277945.1 | NP_001264874.1 | 6 - 6 | 789 | 88 | |
NM_001277946.1 | NP_001264875.1 | 4 - 4 | 574 | 88 | |
NM_001277947.1 | NP_001264876.1 | 6 - 6 | 734 | 88 | |
NM_001277948.1 | NP_001264877.1 | 5 - 5 | 662 | 88 | |
NM_001277949.1 | NP_001264878.1 | 5 - 5 | 701 | 88 | |
NM_001277951.1 | NP_001264880.1 | 3 - 3 | 445 | 88 | |
NM_001277952.1 | NP_001264881.1 | 5 - 5 | 639 | 88 | |
NM_018300.3 | NP_060770.3 | 3 - 3 | 394 | 88 | |
XM_017026951.1 | XP_016882440.1 | 7 - 7 | 891 | 88 | |
GenBank mRNA | AB209516.1 | - | 2 - 2 | 1534 | 88 |
AK001877.1 | - | 1 - 1 | 94 | 88 | |
AK027518.1 | - | 3 - 3 | 439 | 88 | |
AK027782.1 | - | 2 - 2 | 259 | 88 | |
AK055318.1 | - | 2 - 2 | 926 | 88 | |
AK075066.1 | - | 6 - 6 | 696 | 88 | |
AK225945.1 | - | 1 - 1 | 145 | 88 | |
AK289399.1 | - | 4 - 4 | 479 | 88 | |
BC032867.1 | - | 6 - 6 | 718 | 88 | |
BC050407.1 | - | 1 - 1 | 135 | 88 | |
BC063032.1 | - | 2 - 2 | 283 | 88 | |
BX647540.1 | - | 2 - 2 | 2251 | 88 | |
DA107683.1 | - | 1 - 1 | 88 | ||
DA258790.1 | - | 4 - 4 | 476 | 88 | |
DA380756.1 | - | 1 - 1 | 103 | 88 | |
DA731895.1 | - | 3 - 3 | 439 | 88 | |
DR761375.1 | - | 2 - 2 | 332 | 88 |