Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
KRTAP5-4
Entrez Gene ID
387267
Gene Name
keratin associated protein 5-4
Gene Aliases
KRTAP5.4
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012709.1 NP_001012727.1 - 1046 93
GenBank mRNA AB126073.1 - - 1046 93

View Full Product Details