Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
LCE1D
Entrez Gene ID
353134
Gene Name
late cornified envelope 1D
Gene Aliases
LEP4
Chromosome Location
Chr.1: 152796751 - 152798181 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178352.2 NP_848129.1 2 - 2 46 65
GenBank mRNA BG697954.1 - 2 - 2 46 65

View Full Product Details