The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_003529.3 | 5 - 6 | 1128 | 123 | |
NR_047532.1 | 4 - 5 | 815 | 123 | ||
NR_047533.1 | 2 - 3 | 520 | 123 | ||
NR_047534.1 | 2 - 3 | 520 | 123 | ||
NR_047535.1 | 2 - 3 | 520 | 123 | ||
NR_047536.1 | 2 - 3 | 520 | 123 | ||
NR_047537.1 | 2 - 3 | 520 | 123 | ||
NR_047538.1 | 2 - 3 | 520 | 123 | ||
NR_047539.1 | 5 - 6 | 1128 | 123 | ||
NR_047540.1 | 2 - 3 | 520 | 123 | ||
NR_047541.1 | 2 - 3 | 520 | 123 | ||
NR_047542.1 | 2 - 3 | 520 | 123 | ||
NR_047543.1 | 2 - 3 | 520 | 123 | ||
NR_120536.1 | 2 - 3 | 520 | 123 | ||
GenBank mRNA | GQ422823.1 | - | 2 - 3 | 520 | 123 |
GQ495919.1 | - | 2 - 3 | 409 | 123 | |
GQ495920.1 | - | 2 - 3 | 409 | 123 | |
GQ495921.1 | - | 2 - 3 | 409 | 123 | |
GQ495922.1 | - | 2 - 3 | 409 | 123 | |
GQ495923.1 | - | 2 - 3 | 409 | 123 | |
GQ495924.1 | - | 2 - 3 | 409 | 123 | |
GQ495925.1 | - | 2 - 3 | 520 | 123 | |
GQ495926.1 | - | 2 - 3 | 520 | 123 |