Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EFCAB14-AS1
- Entrez Gene ID
-
100130197
- Gene Name
- EFCAB14 antisense RNA 1
- Gene Aliases
-
KIAA0494-AS1
- Chromosome Location
-
Chr.1: 46674036 - 46692098 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_038827.1 |
|
2 - 3 |
275 |
90 |
GenBank mRNA |
BE504902.1 |
- |
2 - 3 |
|
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG195573.1 |
- |
|
93 |
|
Chr. - - [-] on Build |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
testis expressed 38 |
TEX38 |
ATPAF1-AS1, C1orf223, THEG4 |
-8.6kb
|
Sense |
Chr. - 46669344 - 46673603 on Build GRCh38 |
Downstream |
cytochrome P450 family 4 subfamily B member 1 |
CYP4B1 |
CYPIVB1, P-450HP |
112.2kb
|
Sense |
Chr. - 46798995 - 46819349 on Build GRCh38 |
View Full Product Details