The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244638.1 | NP_001231567.1 | 4 - 5 | 644 | 101 |
NM_032199.2 | NP_115575.1 | 7 - 8 | 1560 | 101 | |
XM_011540262.1 | XP_011538564.1 | 6 - 7 | 1329 | 101 | |
GenBank mRNA | AK024803.1 | - | 4 - 5 | 389 | 101 |
BC015120.1 | - | 4 - 5 | 648 | 101 | |
BC066345.1 | - | 4 - 5 | 613 | 101 | |
BC107800.1 | - | 7 - 8 | 1119 | 101 | |
M73837.1 | - | 2 - 3 | 201 | 101 |