Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCAN2
Entrez Gene ID
10231
Gene Name
regulator of calcineurin 2
Gene Aliases
CSP2, DSCR1L1, MCIP2, RCN2, ZAKI-4, ZAKI4
Chromosome Location
Chr.6: 46220730 - 46492067 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251973.1 NP_001238902.1 2 - 3 419 73
NM_001251974.1 NP_001238903.1 2 - 3 372 73
XM_011514226.1 XP_011512528.1 2 - 3 461 73
GenBank mRNA AK090990.1 - 2 - 3 343 73
DA113879.1 - 2 - 3 372 73
DA409014.1 - 1 - 2 127 73
DB289594.1 - 2 - 3 421 73

View Full Product Details