Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VTRNA1-3
Entrez Gene ID
56662
Gene Name
vault RNA 1-3
Gene Aliases
HVG3, VAULTRC3, VR3, hvg-3
Chromosome Location
Chr.5: 140726159 - 140726246 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_026705.1 1 - 1 55 63

View Full Product Details