Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
REREP3
Entrez Gene ID
646396
Gene Name
arginine-glutamic acid dipeptide repeats pseudogene 3
Gene Aliases
-
Chromosome Location
Chr.15: 22258614 - 22282880 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033735.1 1 - 2 262 85
GenBank mRNA DN831801.1 - 1 - 2 262 85
DW010002.1 - 1 - 2 258 85


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DW010003.1 - 1 - 2 Chr. 15 - 22258599 - 22279257 [-] on Build GRCh38 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 1268a MIR1268A MIR1268, MIRN1268, hsa-mir-1268, hsa-mir-1268a -33.3kb Anti-sense Chr. - 22225278 - 22225329 on Build GRCh38
Downstream golgin A8 family member E, pseudogene GOLGA8EP GOLGA8E 145.4kb Anti-sense Chr. - 22424673 - 22438026 on Build GRCh38

View Full Product Details