Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CRIP2
- Entrez Gene ID
-
1397
- Gene Name
- cysteine rich protein 2
- Gene Aliases
-
CRIP, CRP2, ESP1
- Chromosome Location
-
Chr.14: 105472938 - 105480170 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073083.1 |
|
1 - 2 |
135 |
91 |
GenBank mRNA |
CN367966.1 |
- |
1 - 2 |
93 |
91 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
HY171718.1 |
- |
1 - 2 |
107 |
|
Chr. 14 - 105474803 - 105479476 [+] on Build GRCh38 |
91 |
GenBankmRNA |
BM706228.1 |
- |
1 - 2 |
67 |
|
Chr. 14 - 105474843 - 105479757 [+] on Build GRCh38 |
91 |
GenBankmRNA |
BU191847.1 |
- |
|
85 |
|
Chr. - - [-] on Build |
91 |
GenBankmRNA |
BM762285.1 |
- |
1 - 2 |
95 |
|
Chr. 14 - 105474815 - 105479033 [+] on Build GRCh38 |
91 |
GenBankmRNA |
DB490342.1 |
- |
1 - 2 |
92 |
|
Chr. 14 - 105474818 - 105478873 [+] on Build GRCh38 |
91 |
GenBankmRNA |
BQ226901.1 |
- |
|
83 |
|
Chr. - - [-] on Build |
91 |
GenBankmRNA |
BM847123.1 |
- |
1 - 2 |
95 |
|
Chr. 14 - 105474815 - 105479619 [+] on Build GRCh38 |
91 |
GenBankmRNA |
BM008566.1 |
- |
1 - 2 |
86 |
|
Chr. 14 - 105474824 - 105480024 [+] on Build GRCh38 |
91 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
metastasis associated 1 |
MTA1 |
|
-4.0kb
|
Sense |
Chr. - 105419849 - 105470729 on Build GRCh38 |
Downstream |
cysteine rich protein 1 |
CRIP1 |
CRHP, CRIP, CRP-1, CRP1 |
8.4kb
|
Sense |
Chr. - 105486920 - 105488789 on Build GRCh38 |
View Full Product Details