Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SERPINB1
- Entrez Gene ID
-
1992
- Gene Name
- serpin family B member 1
- Gene Aliases
-
EI, ELANH2, HEL-S-27, HEL57, LEI, M/NEI, MNEI, PI-2, PI2
- Chromosome Location
-
Chr.6: 2832332 - 2842049 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073111.1 |
|
2 - 3 |
411 |
99 |
GenBank mRNA |
AK298044.1 |
- |
2 - 3 |
222 |
99 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BP321923.1 |
- |
2 - 3 |
225 |
|
Chr. 6 - 2837915 - 2841863 [-] on Build GRCh38 |
99 |
GenBankmRNA |
BX363740.2 |
- |
2 - 3 |
350 |
|
Chr. 6 - 2836125 - 2841988 [-] on Build GRCh38 |
99 |
GenBankmRNA |
DC358661.1 |
- |
2 - 3 |
222 |
|
Chr. 6 - 2837891 - 2841860 [-] on Build GRCh38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
Werner helicase interacting protein 1 |
WRNIP1 |
WHIP, bA420G6.2 |
-53.6kb
|
Anti-sense |
Chr. - 2765341 - 2785745 on Build GRCh38 |
Downstream |
microRNA 4645 |
MIR4645 |
mir-4645 |
13.4kb
|
Sense |
Chr. - 2854031 - 2854107 on Build GRCh38 |
View Full Product Details