Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NEDD9
- Entrez Gene ID
-
4739
- Gene Name
- neural precursor cell expressed, developmentally down-regulated 9
- Gene Aliases
-
CAS-L, CAS2, CASL, CASS2, HEF1
- Chromosome Location
-
Chr.6: 11183298 - 11382348 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073131.1 |
|
2 - 3 |
370 |
90 |
GenBank mRNA |
BX648041.1 |
- |
2 - 3 |
338 |
90 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD695225.1 |
- |
2 - 3 |
367 |
|
Chr. 6 - 11248363 - 11382316 [-] on Build GRCh38 |
90 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
small integral membrane protein 13 |
SMIM13 |
C6orf228 |
-166.3kb
|
Anti-sense |
Chr. - 11094033 - 11138738 on Build GRCh38 |
Downstream |
uncharacterized LOC105374928 |
LOC105374928 |
|
111.2kb
|
Anti-sense |
Chr. - 11417401 - 11526218 on Build GRCh38 |
View Full Product Details