Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 36281593 - 36347511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136536.4 NP_001130008.2 2 - 3 747 104
NM_001170700.2 NP_001164171.1 2 - 3 902 104
XM_006714014.3 XP_006714077.1 3 - 4 1381 104
XM_011513693.2 XP_011511995.1 3 - 4 1382 104
XM_011513695.2 XP_011511997.1 3 - 4 1381 104
XM_011513696.2 XP_011511998.1 2 - 3 765 104
XM_017008191.1 XP_016863680.1 3 - 4 1382 104
XM_017008192.1 XP_016863681.1 3 - 4 1384 104
XM_017008193.1 XP_016863682.1 3 - 4 1383 104
GenBank mRNA AK131492.1 - 2 - 3 740 104
AK304357.1 - 2 - 3 733 104
AK316523.1 - 2 - 3 733 104
BC157885.1 - 2 - 3 902 104

View Full Product Details