Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DTHD1
Entrez Gene ID
401124
Gene Name
death domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 36281593 - 36347511 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136536.4 NP_001130008.2 4 - 5 1166 89
NM_001170700.2 NP_001164171.1 4 - 5 1321 89
XM_006714014.3 XP_006714077.1 5 - 6 1800 89
GenBank mRNA AK131492.1 - 4 - 5 1159 89
AK304357.1 - 4 - 5 1152 89
AK316523.1 - 4 - 5 1152 89
BC157885.1 - 4 - 5 1321 89

View Full Product Details