The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271006.1 | NP_001257935.1 | 3 - 4 | 193 | 63 |
XM_017024328.1 | XP_016879817.1 | 3 - 4 | 214 | 63 | |
GenBank mRNA | HY110543.1 | - | 3 - 4 | 183 | 63 |