The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270952.1 | NP_001257881.1 | 8 - 9 | 759 | 90 |
NM_006002.4 | NP_005993.1 | 8 - 9 | 681 | 90 | |
XM_011535212.1 | XP_011533514.1 | 9 - 10 | 873 | 90 | |
XM_011535213.2 | XP_011533515.1 | 8 - 9 | 854 | 90 | |
XM_011535214.2 | XP_011533516.1 | 9 - 10 | 941 | 90 | |
XM_017020727.1 | XP_016876216.1 | 7 - 8 | 656 | 90 | |
GenBank mRNA | AK313665.1 | - | 8 - 9 | 671 | 90 |
BC018125.2 | - | 8 - 9 | 640 | 90 | |
BT019359.1 | - | 8 - 9 | 610 | 90 | |
CR456855.1 | - | 8 - 9 | 610 | 90 | |
M30496.1 | - | 8 - 9 | 630 | 90 |