Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL1RL1
Entrez Gene ID
9173
Gene Name
interleukin 1 receptor like 1
Gene Aliases
DER4, FIT-1, IL33R, ST2, ST2L, ST2V, T1
Chromosome Location
Chr.2: 102311502 - 102352367 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282408.1 NP_001269337.1 7 - 7 883 99
NM_003856.3 NP_003847.2 8 - 8 1203 99
NR_104167.1 9 - 9 1373 99
XM_011512151.1 XP_011510453.1 8 - 8 1210 99
GenBank mRNA AB029084.1 - 9 - 9 1335 99
AK291578.1 - 8 - 8 1195 99
AK303389.1 - 7 - 7 822 99
BC030975.1 - 8 - 8 1196 99
D12763.1 - 7 - 7 1012 99

View Full Product Details