Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX091769.1 |
- |
1 - 2 |
250 |
|
Chr. 8 - 38700663 - 38704855 [-] on Build GRCh38 |
108 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
ring finger protein 5 pseudogene 1 |
RNF5P1 |
|
-100.3kb
|
Sense |
Chr. - 38600173 - 38601257 on Build GRCh38 |
Downstream |
transforming acidic coiled-coil containing protein 1 |
TACC1 |
Ga55 |
23.3kb
|
Anti-sense |
Chr. - 38728186 - 38853028 on Build GRCh38 |
View Full Product Details