The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000431.3 | NP_000422.1 | 6 - 7 | 825 | 84 |
NM_001114185.2 | NP_001107657.1 | 6 - 7 | 816 | 84 | |
NM_001301182.1 | NP_001288111.1 | 5 - 6 | 669 | 84 | |
XM_011538372.2 | XP_011536674.2 | 6 - 7 | 933 | 84 | |
XM_017019313.1 | XP_016874802.1 | 5 - 6 | 622 | 84 | |
XM_017019314.1 | XP_016874803.1 | 6 - 7 | 1291 | 84 | |
GenBank mRNA | AB209722.1 | - | 6 - 7 | 708 | 84 |
AK293130.1 | - | 4 - 5 | 392 | 84 | |
AK295338.1 | - | 5 - 6 | 527 | 84 | |
AK315678.1 | - | 6 - 7 | 704 | 84 | |
BC016140.1 | - | 6 - 7 | 668 | 84 | |
KU178205.1 | - | 5 - 6 | 632 | 84 | |
KU178206.1 | - | 4 - 5 | 476 | 84 | |
M88468.1 | - | 6 - 7 | 723 | 84 |