The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000431.3 | NP_000422.1 | 8 - 9 | 965 | 89 |
NM_001114185.2 | NP_001107657.1 | 8 - 9 | 956 | 89 | |
NM_001301182.1 | NP_001288111.1 | 7 - 8 | 809 | 89 | |
XM_011538372.2 | XP_011536674.2 | 8 - 9 | 1073 | 89 | |
XM_017019313.1 | XP_016874802.1 | 7 - 8 | 762 | 89 | |
XM_017019314.1 | XP_016874803.1 | 8 - 9 | 1431 | 89 | |
GenBank mRNA | AB209722.1 | - | 8 - 9 | 848 | 89 |
AK023087.1 | - | 1 - 2 | 1301 | 89 | |
AK293130.1 | - | 6 - 7 | 532 | 89 | |
AK295338.1 | - | 7 - 8 | 667 | 89 | |
AK315678.1 | - | 8 - 9 | 844 | 89 | |
BC016140.1 | - | 8 - 9 | 808 | 89 | |
KU178205.1 | - | 7 - 8 | 772 | 89 | |
KU178206.1 | - | 6 - 7 | 616 | 89 | |
M88468.1 | - | 8 - 9 | 863 | 89 |