Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MT1F
Entrez Gene ID
4494
Gene Name
metallothionein 1F
Gene Aliases
MT1
Chromosome Location
Chr.16: 56657943 - 56659303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001301272.1 NP_001288201.1 2 - 3 253 166

View Full Product Details