The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301833.1 | NP_001288762.1 | 4 - 5 | 346 | 94 |
NM_001301852.1 | NP_001288781.1 | 3 - 4 | 268 | 94 | |
NM_015955.3 | NP_057039.1 | 3 - 4 | 635 | 94 | |
NR_126032.1 | 3 - 4 | 635 | 94 | ||
NR_126034.1 | 3 - 4 | 268 | 94 | ||
XM_011532892.2 | XP_011531194.1 | 4 - 5 | 283 | 94 | |
XM_011532896.2 | XP_011531198.1 | 2 - 3 | 161 | 94 | |
XM_017004255.1 | XP_016859744.1 | 4 - 5 | 256 | 94 | |
GenBank mRNA | AB041018.1 | - | 3 - 4 | 635 | 94 |
AB041019.1 | - | 3 - 4 | 284 | 94 | |
AF132961.1 | - | 3 - 4 | 333 | 94 | |
AF363446.1 | - | 3 - 4 | 552 | 94 | |
AF529368.1 | - | 3 - 4 | 218 | 94 | |
AK290753.1 | - | 4 - 5 | 302 | 94 | |
BC018733.1 | - | 3 - 4 | 600 | 94 | |
BC070046.1 | - | 4 - 5 | 375 | 94 | |
BC094681.1 | - | 3 - 4 | 239 | 94 | |
BG033287.1 | - | 3 - 4 | 217 | 94 | |
BP248979.1 | - | 4 - 5 | 346 | 94 | |
CD250945.1 | - | 3 - 4 | 332 | 94 |