The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005157.5 | NP_005148.2 | 9 - 10 | 1697 | 67 |
NM_007313.2 | NP_009297.2 | 9 - 10 | 2001 | 67 | |
GenBank mRNA | AB209456.1 | - | 9 - 10 | 1707 | 67 |
AB209642.1 | - | 9 - 10 | 1929 | 67 | |
BC117451.1 | - | 9 - 10 | 1627 | 67 | |
M14752.1 | - | 9 - 10 | 1869 | 67 | |
X16416.1 | - | 9 - 10 | 1652 | 67 |